Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MED16 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_005481.2 | NP_005472.2 | 7 | 1158 | ||
XM_017026120.1 | XP_016881609.1 | 7 | 1153 | |||
XM_017026121.1 | XP_016881610.1 | 6 | 998 | |||
GenBank mRNA | AF106934.1 | - | 7 | 1145 | ||
AF121228.1 | - | 6 | 1007 | |||
BC004554.1 | - | 7 | 1139 | 4053743 | ||
BC007853.2 | - | 3 | 268 | |||
BC011841.1 | - | 7 | 1139 | 4053664 | ||
BC017282.2 | - | 7 | 1133 | 4876977 | ||
EU392449.1 | - | 6 | 1024 | |||
EU392451.1 | - | 5 | 586 |