Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SLC35A2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001032289.2 | NP_001027460.1 | 3 | 618 | ||
NM_001042498.2 | NP_001035963.1 | 3 | 618 | |||
NM_001282647.1 | NP_001269576.1 | 3 | 618 | |||
NM_001282648.1 | NP_001269577.1 | 3 | 305 | |||
NM_001282649.1 | NP_001269578.1 | 2 | 435 | |||
NM_001282650.1 | NP_001269579.1 | 3 | 657 | |||
NM_001282651.1 | NP_001269580.1 | 4 | 702 | |||
NM_005660.2 | NP_005651.1 | 3 | 618 | |||
GenBank mRNA | AK090730.1 | - | 2 | 90 | ||
AK225508.1 | - | 3 | 306 | |||
AK290284.1 | - | 3 | 306 | |||
AK292816.1 | - | 3 | 335 | |||
AK293415.1 | - | 2 | 123 | |||
AK293419.1 | - | 3 | 345 | |||
AK298484.1 | - | 4 | 390 | |||
AK299748.1 | - | 3 | 345 | |||
AL556714.3 | - | 3 | 305 | |||
BC035747.1 | - | 3 | 316 | 5749525 | ||
BI820134.1 | - | 3 | 289 | |||
D84454.1 | - | 3 | 619 | |||
D88146.1 | - | 3 | 300 |