Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ZNF799 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001080821.2 | NP_001074290.1 | 4 | 694 | ||
NM_001322497.1 | NP_001309426.1 | 4 | 1144 | |||
NM_001322498.1 | NP_001309427.1 | 5 | 812 | |||
XM_017027452.1 | XP_016882941.1 | 3 | 487 | |||
GenBank mRNA | AK027660.1 | - | 4 | 1144 | ||
AL556986.3 | - | 4 | 624 | |||
AL832890.1 | - | 3 | 2184 |
Gene Symbol: ZNF443 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_005815.4 | NP_005806.2 | 4 | 688 | ||
XM_011527622.2 | XP_011525924.1 | 4 | 1215 | |||
XM_011527623.2 | XP_011525925.2 | 4 | 1756 | |||
XM_017026139.1 | XP_016881628.1 | 4 | 1756 | |||
XM_017026140.1 | XP_016881629.1 | 3 | 2259 | |||
GenBank mRNA | AB011414.1 | - | 4 | 594 | ||
AK023025.1 | - | 4 | 570 | |||
AK299532.1 | - | 4 | 525 | |||
BC032753.1 | - | 4 | 592 | 5583971 |