Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MYL12A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001303047.1 | NP_001289976.1 | 5 | 1086 | ||
NM_001303048.1 | NP_001289977.1 | 4 | 729 | |||
NM_001303049.1 | NP_001289978.1 | 4 | 736 | |||
NM_006471.3 | NP_006462.1 | 4 | 1003 | |||
GenBank mRNA | AK222569.1 | - | 4 | 679 | ||
AK291145.1 | - | 4 | 699 | |||
BC016372.1 | - | 4 | 678 | 4293343 | ||
BC031972.1 | - | 4 | 1003 | 4836883 | ||
BC032748.1 | - | 4 | 674 | 5563221 | ||
BM561281.1 | - | 5 | 803 | |||
D82059.1 | - | 4 | 669 | |||
EU794621.1 | - | 4 | 1003 |