Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: UGP2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001001521.1 | NP_001001521.1 | 2 | 349 | ||
NM_006759.3 | NP_006750.3 | 2 | 405 | |||
XM_005264537.2 | XP_005264594.1 | 3 | 457 | |||
XM_005264538.1 | XP_005264595.1 | 3 | 424 | |||
XM_006712088.1 | XP_006712151.1 | 2 | 361 | |||
XM_017004855.1 | XP_016860344.1 | 2 | 337 | |||
XM_017004856.1 | XP_016860345.1 | 3 | 229 | |||
XM_017004857.1 | XP_016860346.1 | 2 | 361 | |||
GenBank mRNA | AF086113.1 | - | 2 | 183 | ||
AK300731.1 | - | 2 | 133 | |||
AK314263.1 | - | 2 | 396 | |||
BC000173.2 | - | 2 | 350 | 2985723 | ||
BC002954.1 | - | 2 | 229 | 3542849 | ||
BC047004.1 | - | 2 | 356 | 3924393 | ||
CA389301.1 | - | 2 | 407 | |||
CD516624.1 | - | 2 | 353 | |||
U27460.1 | - | 3 | 174 |