Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: LILRA3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001172654.2 | NP_001166125.1 | 6 | 584 | ||
NM_006865.4 | NP_006856.3 | 6 | 820 | |||
GenBank mRNA | AF014923.1 | - | 6 | 728 | ||
AF014924.1 | - | 6 | 536 | |||
AF025527.1 | - | 6 | 821 | |||
BC028208.1 | - | 6 | 793 | 5215423 | ||
U91926.1 | - | 6 | 790 | |||
U91927.1 | - | 4 | 671 |