Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: RASGRF2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_006909.2 | NP_008840.1 | 27 | 3997 | ||
XM_017009682.1 | XP_016865171.1 | 27 | 3679 | |||
GenBank mRNA | AL598102.1 | - | 3 | 475 | ||
CR749239.1 | - | 5 | 2019 | |||
DA219722.1 | - | 2 | 345 |