Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: RNF5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_006913.3 | NP_008844.1 | 6 | 1127 | ||
GenBank mRNA | BC004155.2 | - | 1 | 1004 | 2822537 | |
BC020290.1 | - | 5 | 861 | |||
BC111392.1 | - | 6 | 1121 | 6458319 |