Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SLC35D2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001286990.1 | NP_001273919.1 | 3 | 326 | ||
NM_007001.2 | NP_008932.2 | 3 | 326 | |||
NR_104627.1 | 3 | 326 | ||||
XM_005251673.1 | XP_005251730.1 | 3 | 301 | |||
XM_005251674.1 | XP_005251731.1 | 3 | 301 | |||
XM_005251675.2 | XP_005251732.1 | 3 | 311 | |||
XM_005251676.1 | XP_005251733.1 | 3 | 301 | |||
XM_005251678.4 | XP_005251735.1 | 1 | 33 | |||
XM_006716939.2 | XP_006717002.1 | 3 | 311 | |||
XM_011518164.1 | XP_011516466.1 | 3 | 311 | |||
GenBank mRNA | AB106537.1 | - | 3 | 259 | ||
AB122077.1 | - | 3 | 271 | |||
AJ005866.1 | - | 1 | 22 | |||
AK307890.1 | - | 3 | 305 | |||
BC100278.1 | - | 3 | 317 | 6056863 | ||
BC113579.1 | - | 4 | 279 | 8322631 | ||
BX361048.2 | - | 3 | 325 |