Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CD300A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001256841.1 | NP_001243770.1 | 2 | 420 | ||
NM_007261.3 | NP_009192.2 | 3 | 759 | |||
XM_005256990.2 | XP_005257047.1 | 3 | 780 | |||
XM_005256991.4 | XP_005257048.1 | 3 | 782 | |||
XM_005256992.4 | XP_005257049.1 | 3 | 688 | |||
XM_006721656.3 | XP_006721719.1 | 3 | 796 | |||
XM_011524254.2 | XP_011522556.1 | 2 | 483 | |||
GenBank mRNA | AF020314.1 | - | 3 | 515 | ||
AF161346.1 | - | 2 | 146 | |||
AJ010101.1 | - | 3 | 467 | |||
AJ010102.1 | - | 2 | 128 | |||
AJ224864.1 | - | 3 | 740 | |||
AJ238323.1 | - | 3 | 467 | |||
AK309631.1 | - | 4 | 824 | |||
AK309636.1 | - | 3 | 466 | |||
BC032352.1 | - | 3 | 724 | 5218501 |