Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: KLRC3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_002261.2 | NP_002252.2 | 6 | 665 | ||
NM_007333.2 | NP_031359.2 | 6 | 665 | |||
GenBank mRNA | AF078550.1 | - | 6 | 620 | ||
AF350016.1 | - | 6 | 620 | |||
AF350017.1 | - | 6 | 620 | |||
AF461157.1 | - | 6 | 636 | |||
L14542.1 | - | 6 | 665 |