Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: DTYMK | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001165031.1 | NP_001158503.1 | 4 | 616 | ||
NM_001320902.1 | NP_001307831.1 | 3 | 559 | |||
NM_001320903.1 | NP_001307832.1 | 3 | 525 | |||
NM_001320904.1 | NP_001307833.1 | 4 | 631 | |||
NM_001320905.1 | NP_001307834.1 | 5 | 805 | |||
NM_012145.3 | NP_036277.2 | 4 | 688 | |||
NR_033255.1 | 3 | 597 | ||||
NR_135492.1 | 4 | 650 | ||||
GenBank mRNA | AK223434.1 | - | 4 | 535 | ||
BC001827.2 | - | 4 | 528 | 3533327 | ||
BC147006.1 | - | 4 | 627 | 9056934 | ||
BC147010.1 | - | 4 | 627 | 9056938 | ||
BC171894.1 | - | 4 | 623 | |||
BC171902.1 | - | 4 | 555 | 9054556 | ||
BC171903.1 | - | 3 | 536 | |||
BG765702.1 | - | 3 | 343 | |||
BM552707.1 | - | 4 | 674 | |||
CN356277.1 | - | 3 | 395 | |||
CR541986.1 | - | 4 | 503 | |||
CR542015.1 | - | 4 | 503 | |||
CX166352.1 | - | 4 | 688 | |||
L16991.1 | - | 4 | 545 | |||
X54729.1 | - | 4 | 527 |