Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TIMM10 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_012456.2 | NP_036588.1 | 3 | 344 | ||
GenBank mRNA | AF150089.1 | - | 3 | 326 | ||
AF152354.1 | - | 2 | 197 | |||
AK289751.1 | - | 3 | 295 | |||
BC032133.2 | - | 3 | 307 | 4761702 | ||
CB106298.1 | - | 3 | 344 |