Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PYCARD | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_013258.4 | NP_037390.2 | 3 | 726 | ||
NM_145182.2 | NP_660183.1 | 2 | 669 | |||
GenBank mRNA | AB023416.2 | - | 3 | 726 | ||
AF184073.1 | - | 3 | 714 | |||
AF255794.1 | - | 2 | 657 | |||
AF310103.1 | - | 3 | 640 | |||
AF384665.1 | - | 3 | 693 | |||
AK000211.1 | - | 3 | 678 | |||
BC004470.2 | - | 4 | 696 | 3678925 | ||
BC013569.1 | - | 3 | 633 |