Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MYLPF | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001324458.1 | NP_001311387.1 | 7, 8 | 567 | ||
NM_001324459.1 | NP_001311388.1 | 7, 8 | 595 | |||
NM_013292.4 | NP_037424.2 | 5, 6 | 430 | |||
GenBank mRNA | AF363061.1 | - | 5, 6 | 403 | ||
AK314082.1 | - | 5, 6 | 430 | |||
BC012571.1 | - | 5, 6 | 408 | 4245958 | ||
BX649054.1 | - | 5, 6 | 398 | |||
CR456963.1 | - | 4, 5 | 349 | |||
M21812.1 | - | 5, 6 | 410 | |||
X57543.1 | - | 5, 6 | 410 |