Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ANAPC2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_013366.3 | NP_037498.1 | 2694 | |||
GenBank mRNA | BC001579.1 | - | 1886 | |||
BC032503.1 | - | 2626 | 5243107 |