Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: REM1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_014012.5 | NP_054731.2 | 1664 | |||
GenBank mRNA | AF152863.1 | - | 1646 |