Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PDSS1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001321978.1 | NP_001308907.1 | 5 | 735 | ||
NM_001321979.1 | NP_001308908.1 | 5 | 735 | |||
NM_014317.4 | NP_055132.2 | 5 | 735 | |||
XM_011519437.2 | XP_011517739.1 | 2 | 1135 | |||
XM_017016011.1 | XP_016871500.1 | 3 | 213 | |||
GenBank mRNA | AB209763.1 | - | 4 | 2773 | ||
AB210838.1 | - | 5 | 493 | |||
AF118395.1 | - | 4 | 355 | |||
AK024802.1 | - | 5 | 500 | |||
AK223414.1 | - | 5 | 500 | |||
BC049211.1 | - | 5 | 445 | |||
BC063635.1 | - | 5 | 463 | 4397753 |