Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ENPP4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_014936.4 | NP_055751.1 | 4 | 2270 | ||
GenBank mRNA | AB020686.1 | - | 3 | 2059 | ||
AJ420515.1 | - | 1 | 37 |