Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MAST1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_014975.2 | NP_055790.1 | 14 | 1710 | ||
XM_011527805.2 | XP_011526107.1 | 15 | 1607 | |||
XM_011527808.2 | XP_011526110.1 | 3 | 549 | |||
XM_017026501.1 | XP_016881990.1 | 11 | 1163 | |||
GenBank mRNA | AB023190.1 | - | 14 | 1621 | ||
AK297553.1 | - | 14 | 1614 | |||
BC027985.1 | - | 3 | 428 |