Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TBC1D9 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_015130.2 | NP_055945.2 | 7 | 1510 | ||
GenBank mRNA | AB020689.2 | - | 7 | 1313 | ||
AB449892.1 | - | 7 | 1238 | |||
AK127020.1 | - | 4 | 798 | |||
BC146758.1 | - | 7 | 1313 | 9007196 | ||
CN332501.1 | - | 2 | 257 |