Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: LARP4B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_015155.2 | NP_055970.1 | 3 | 420 | ||
XM_005252432.3 | XP_005252489.1 | 3 | 429 | |||
XM_005252435.3 | XP_005252492.1 | 2 | 142 | |||
XM_011519435.2 | XP_011517737.1 | 3 | 264 | |||
XM_017015990.1 | XP_016871479.1 | 3 | 1988 | |||
XM_017015993.1 | XP_016871482.1 | 2 | 148 | |||
XM_017015994.1 | XP_016871483.1 | 3 | 185 | |||
GenBank mRNA | BC131630.1 | - | 2 | 137 | 40118844 | |
BC152443.1 | - | 2 | 144 | 8862568 | ||
D86971.2 | - | 2 | 144 | |||
HY013410.1 | - | 3 | 420 |