Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CUX2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_015267.3 | NP_056082.2 | 6, 7 | 708 | ||
XM_011538061.2 | XP_011536363.1 | 6, 7 | 2218 | |||
XM_011538063.2 | XP_011536365.1 | 5, 6 | 2083 | |||
XM_011538069.2 | XP_011536371.1 | 6, 7 | 557 | |||
XM_011538070.2 | XP_011536372.1 | 6, 7 | 1467 | |||
XM_017019080.1 | XP_016874569.1 | 7, 8 | 1550 | |||
XM_017019081.1 | XP_016874570.1 | 7, 8 | 613 | |||
GenBank mRNA | AB006631.2 | - | 6, 7 | 707 | ||
AF234996.1 | - | 7 | 871 | |||
AF271236.1 | - | 6 | 618 | |||
BC151245.1 | - | 6, 7 | 718 | 8860519 |