Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: DNAJC16 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001287811.1 | NP_001274740.1 | 14 | 2233 | ||
NM_015291.3 | NP_056106.1 | 15 | 2418 | |||
NR_109898.1 | 15 | 2648 | ||||
GenBank mRNA | AB023179.2 | - | 15 | 2374 | ||
AK022530.1 | - | 6 | 785 | |||
AK023805.1 | - | 11 | 1588 | |||
AK292479.1 | - | 15 | 2350 | |||
AL137715.1 | - | 10 | 1850 | |||
AL833299.1 | - | 15 | 2587 | |||
BC031991.1 | - | 7 | 968 | |||
BC047363.1 | - | 15 | 2418 | 5269539 | ||
CR749259.1 | - | 14 | 2178 | |||
CR749567.1 | - | 14 | 2178 |