Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SRGAP2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001170637.3 | NP_001164108.1 | 22 | 5275 | ||
NM_015326.4 | NP_056141.2 | 22 | 5278 | |||
XM_005277510.2 | XP_005277567.1 | 23 | 5465 | |||
XM_005277511.2 | XP_005277568.1 | 23 | 5462 | |||
XM_011509354.1 | XP_011507656.1 | 24 | 5504 | |||
XM_011509355.1 | XP_011507657.1 | 24 | 5501 | |||
XM_011509356.2 | XP_011507658.1 | 25 | 5599 | |||
XM_017000838.1 | XP_016856327.1 | 22 | 5148 | |||
XM_017000839.1 | XP_016856328.1 | 22 | 5178 | |||
XM_017000840.1 | XP_016856329.1 | 24 | 4793 | |||
GenBank mRNA | AB007925.1 | - | 22 | 4810 |