Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
Integrated CRO-CDMO Services
CDMO Services
CRO Services
Custom Services
Enterprise Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Thermo Fisher Scientific
Search All
Search
Search button
Search button
Close
Order Status
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other ERC2 SIRNA Assays ›
Gene Symbol:
ERC2
Gene Name:
ELKS/RAB6-interacting/CAST family member 2
Gene Aliases:
CAST, CAST1, ELKSL, SPBC110, Spc110
Chromosome Location:
Chr.3: 55508311 - 56468473 on Build GRCh38
UniGene ID:
Hs.476389
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID
289749
Purification
HPLC
Standard
In-Vivo Ready
Size
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
Availability
Inventoried
Catalog #
AM16708
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: ERC2
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_015576.2
NP_056391.1
10
2292
NR_132749.1
10
2292
XM_017006138.1
XP_016861627.1
12
2372
XM_017006139.1
XP_016861628.1
11
2330
XM_017006140.1
XP_016861629.1
11
2329
XM_017006141.1
XP_016861630.1
10
2320
XM_017006142.1
XP_016861631.1
10
2320
XM_017006143.1
XP_016861632.1
11
2329
XM_017006144.1
XP_016861633.1
11
2329
XM_017006145.1
XP_016861634.1
11
2329
XM_017006146.1
XP_016861635.1
10
2320
XM_017006147.1
XP_016861636.1
11
2329
XM_017006148.1
XP_016861637.1
10
2320
XM_017006149.1
XP_016861638.1
10
2379
XM_017006150.1
XP_016861639.1
10
2320
XM_017006151.1
XP_016861640.1
10
2320
XM_017006152.1
XP_016861641.1
11
2329
XM_017006153.1
XP_016861642.1
11
2329
XM_017006154.1
XP_016861643.1
11
2329
XM_017006155.1
XP_016861644.1
10
2402
XM_017006156.1
XP_016861645.1
10
2320
XM_017006157.1
XP_016861646.1
11
2329
XM_017006158.1
XP_016861647.1
10
2320
XM_017006159.1
XP_016861648.1
10
2359
GenBank mRNA
AB002376.2
-
10
2292
BC111550.1
-
10
2266
40008953
BC112391.1
-
10
2266
40008962
Back To Top
More Information
Panther Classification:
Molecular Function -
membrane traffic protein
Gene Ontology Categories:
Process(es)
protein binding
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-blue-65d5d77f49-g5d4h:80/100.66.76.71:80.
git-commit: a4c025c8ba47546866237b65de1f276a5fdbbe78
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.18.1-Offline