Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PITPNM2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001300801.1 | NP_001287730.1 | 25 | 4854 | ||
NM_020845.2 | NP_065896.1 | 25 | 4811 | |||
XM_011538590.2 | XP_011536892.1 | 26 | 5133 | |||
GenBank mRNA | AB040890.2 | - | 26 | 4875 | ||
AF334585.1 | - | 24 | 4750 | |||
AL133612.1 | - | 16 | 3258 | |||
BC050567.1 | - | 1 | 42 |