Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TPRA1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001136053.2 | NP_001129525.1 | 7 | 913 | ||
NM_001142646.2 | NP_001136118.1 | 7 | 913 | |||
NR_073377.1 | 7 | 913 | ||||
XM_006713495.3 | XP_006713558.1 | 8 | 1094 | |||
XM_006713496.2 | XP_006713559.1 | 7 | 771 | |||
XM_006713497.3 | XP_006713560.1 | 8 | 1138 | |||
XM_011512422.1 | XP_011510724.1 | 7 | 687 | |||
XM_017005707.1 | XP_016861196.1 | 8 | 877 | |||
XM_017005708.1 | XP_016861197.1 | 8 | 907 | |||
XM_017005709.1 | XP_016861198.1 | 8 | 1008 | |||
GenBank mRNA | AB037108.1 | - | 6 | 837 | ||
AF289568.1 | - | 7 | 860 | |||
BC017058.1 | - | 7 | 682 | 3904719 | ||
BC050711.2 | - | 7 | 807 | 6168632 | ||
CD580484.2 | - | 7 | 648 |