Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MAPK8IP2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_012324.4 | NP_036456.1 | 12 | 3191 | ||
XM_011530679.2 | XP_011528981.1 | 12 | 3083 | |||
XM_011530680.2 | XP_011528982.1 | 12 | 2969 | |||
XM_011530681.2 | XP_011528983.1 | 12 | 2948 | |||
GenBank mRNA | AF136382.1 | - | 12 | 3174 | ||
AL021708.1 | - | 8 | 2194 | |||
BC009940.2 | - | 11 | 1941 | 4100637 | ||
BC047527.1 | - | 12 | 3103 | 5277237 | ||
U79261.1 | - | 7 | 1265 |