Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: VRK3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001025778.1 | NP_001020949.1 | 14 | 1924 | ||
NM_016440.3 | NP_057524.3 | 15 | 2074 | |||
XM_005258971.3 | XP_005259028.1 | 15 | 2401 | |||
XM_005258972.3 | XP_005259029.1 | 14 | 2251 | |||
XM_006723237.3 | XP_006723300.1 | 15 | 2215 | |||
XM_006723238.3 | XP_006723301.1 | 14 | 1865 | |||
GenBank mRNA | AV706241.1 | - | 2 | 331 |