Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CPSF2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001322270.1 | NP_001309199.1 | 9 | 1372 | ||
NM_001322271.1 | NP_001309200.1 | 8 | 1212 | |||
NM_001322272.1 | NP_001309201.1 | 9 | 1577 | |||
NM_017437.2 | NP_059133.1 | 9 | 1372 | |||
GenBank mRNA | AB037788.1 | - | 3 | 496 | ||
AK001627.2 | - | 9 | 1349 | |||
AK001912.1 | - | 7 | 928 | |||
AK023583.1 | - | 6 | 949 | |||
AL442079.1 | - | 2 | 271 | |||
BC070095.1 | - | 9 | 1342 | 5267724 |