Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PCSK4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_017573.4 | NP_060043.2 | 5 | 649 | ||
XM_005259586.1 | XP_005259643.1 | 5 | 491 | |||
XM_011528085.2 | XP_011526387.1 | 6 | 874 | |||
XM_011528086.2 | XP_011526388.1 | 6 | 874 | |||
XM_011528087.2 | XP_011526389.1 | 6 | 874 | |||
XM_011528088.2 | XP_011526390.1 | 6 | 526 | |||
XM_011528089.2 | XP_011526391.1 | 6 | 874 | |||
XM_011528090.2 | XP_011526392.1 | 5 | 793 | |||
XM_011528091.2 | XP_011526393.1 | 4 | 347 | |||
XM_011528092.2 | XP_011526394.1 | 6 | 874 | |||
XM_011528093.2 | XP_011526395.1 | 6 | 874 | |||
XM_011528094.1 | XP_011526396.1 | 4 | 570 | |||
XM_011528095.2 | XP_011526397.1 | 6 | 874 | |||
XM_011528096.1 | XP_011526398.1 | 5 | 491 | |||
XM_017026897.1 | XP_016882386.1 | 3 | 251 | |||
GenBank mRNA | AK057235.1 | - | 4 | 801 | ||
AK302079.1 | - | 5 | 604 | |||
AY358963.1 | - | 5 | 607 | |||
BC142942.1 | - | 4 | 870 |