Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ZNF446 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001304453.1 | NP_001291382.1 | 4 | 723 | ||
NM_017908.3 | NP_060378.1 | 5 | 1064 | |||
XM_005259052.4 | XP_005259109.1 | 5 | 1065 | |||
XM_005259053.4 | XP_005259110.1 | 5 | 1065 | |||
XM_006723266.3 | XP_006723329.1 | 5 | 1065 | |||
XM_017026949.1 | XP_016882438.1 | 5 | 1065 | |||
XM_017026950.1 | XP_016882439.1 | 5 | 1065 | |||
GenBank mRNA | AK000633.1 | - | 5 | 803 | ||
AK026153.1 | - | 5 | 778 | |||
AL122108.1 | - | 1 | 46 | |||
AL833955.1 | - | 4 | 734 | |||
AY280800.1 | - | 5 | 843 | |||
BC017206.2 | - | 5 | 785 | 3636172 | ||
BQ441187.1 | - | 4 | 450 |