Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ATAD3A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001170535.1 | NP_001164006.1 | 16 | 2251 | ||
NM_001170536.1 | NP_001164007.1 | 16 | 2081 | |||
NM_018188.3 | NP_060658.3 | 16 | 2395 | |||
GenBank mRNA | AK001571.1 | - | 16 | 2234 | ||
AK025865.1 | - | 9 | 1251 | |||
AK056099.1 | - | 16 | 2081 | |||
AK091144.1 | - | 16 | 2363 | |||
AK092833.1 | - | 1 | 2367 | |||
BC007803.2 | - | 16 | 2212 | 4136170 | ||
BC011814.2 | - | 16 | 2217 | 3162989 | ||
BC014101.2 | - | 16 | 2241 | 3503438 | ||
BC033109.1 | - | 16 | 2215 | 4877449 | ||
BC063607.1 | - | 16 | 2209 | 4939339 |