Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
Integrated CRO-CDMO Services
CDMO Services
CRO Services
Custom Services
Enterprise Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Thermo Fisher Scientific
Search All
Search
Search button
Search button
Close
Order Status
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other EEF1A1 SIRNA Assays ›
Gene Symbol:
EEF1A1
Gene Name:
eukaryotic translation elongation factor 1 alpha 1
Gene Aliases:
CCS-3, CCS3, EE1A1, EEF-1, EEF1A, EF-Tu, EF1A, GRAF-1EF, HNGC:16303, LENG7, PTI1, eEF1A-1
Chromosome Location:
Chr.6: 73515750 - 73521032 on Build GRCh38
UniGene ID:
Hs.535192
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID
2898
Purification
HPLC
Standard
In-Vivo Ready
Size
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
Availability
Inventoried
Catalog #
AM16708
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: EEF1A1
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_001402.5
NP_001393.1
5
707
XM_011535514.2
XP_011533816.1
5
1342
GenBank mRNA
AF174496.1
-
4
537
AF328730.1
-
1
415
AF397403.1
-
3
452
AJ420488.1
-
4
666
AK024888.1
-
1
705
AK026650.1
-
1
707
AK055058.1
-
5
707
AK058051.1
-
5
707
AK058163.1
-
4
525
AK098510.1
-
5
707
AK098518.1
-
5
567
AK098587.1
-
2
135
AK098623.1
-
5
707
AK127384.1
-
5
707
AK222515.1
-
1
707
AK222519.1
-
1
356
AK222523.1
-
1
707
AK222982.1
-
1
707
AK223030.1
-
1
704
AK223042.1
-
1
562
AK223046.1
-
1
710
AK289374.1
-
5
707
AK289442.1
-
5
645
AK290691.1
-
5
707
AK292639.1
-
5
1023
AK297878.1
-
6
505
AK300922.1
-
5
568
AK304214.1
-
6
667
AK315827.1
-
5
698
AY043301.1
-
4
1054
BC008587.1
-
1
693
3447654
BC009733.1
-
1
657
3877516
BC009875.2
-
1
697
3948601
BC010735.1
-
1
686
3898364
BC012891.1
-
1
685
3858483
BC014224.2
-
1
696
4335756
BC014377.1
-
1
119
BC018150.1
-
1
693
3851939
BC018641.2
-
1
693
3660140
BC018850.2
-
1
696
BC019050.2
-
1
659
BC020477.2
-
1
356
BC021686.1
-
1
712
3930863
BC028674.1
-
1
711
4107346
BC029337.1
-
1
681
BC029343.1
-
1
679
BC029997.1
-
1
694
BC035877.1
-
1
708
BC038339.1
-
1
693
3461588
BC057391.1
-
1
662
4339606
BC063511.1
-
1
130
BC066893.1
-
1
707
4818863
BC071619.1
-
2
623
4448926
BC071727.1
-
1
662
4514479
BC071741.1
-
1
687
3885539
BC071841.1
-
1
682
4587981
BC072385.1
-
1
694
5585092
BC082268.1
-
1
686
6729595
BC094687.1
-
2
599
30366186
BC111051.1
-
1
697
5585079
DQ867534.1
-
1
393
EF219140.1
-
5
867
EF362804.1
-
5
937
M29548.1
-
2
239
X03558.1
-
5
697
X16869.1
-
4
659
Back To Top
More Information
Panther Classification:
Molecular Function -
translation factor
Gene Ontology Categories:
Function(s)
Process(es)
transcription, DNA-templated
regulation of transcription, DNA-templated
translational elongation
cellular response to epidermal growth factor stimulus
regulation of chaperone-mediated autophagy
tRNA binding
translation elongation factor activity
GTPase activity
protein binding
GTP binding
protein kinase binding
poly(A) RNA binding
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-green-cccc65c97-6lbnp:80/10.239.161.162:80.
git-commit: 2b810224bbfb944e11adb8d8c7a69f36ca15064f
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.16.1-offline