Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ZKSCAN7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001288590.1 | NP_001275519.1 | 3 | 872 | ||
NM_001288591.1 | NP_001275520.1 | 3 | 872 | |||
NM_001288592.1 | NP_001275521.1 | 2 | 331 | |||
NM_018651.3 | NP_061121.2 | 3 | 909 | |||
NM_025169.2 | NP_079445.1 | 3 | 909 | |||
GenBank mRNA | AF154846.1 | - | 2 | 525 | ||
AK022800.1 | - | 3 | 818 | |||
AK291285.1 | - | 3 | 854 | |||
AY280798.1 | - | 3 | 863 | |||
BC010555.1 | - | 3 | 827 | |||
BC063020.1 | - | 3 | 861 | |||
BC120982.1 | - | 3 | 601 | |||
BC120983.1 | - | 3 | 640 | |||
BC127639.1 | - | 3 | 640 | |||
BC127640.1 | - | 3 | 640 | |||
BC148260.1 | - | 2 | 62 |