Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
Integrated CRO-CDMO Services
CDMO Services
CRO Services
Custom Services
Enterprise Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Thermo Fisher Scientific
Search All
Search
Search button
Search button
Close
Order Status
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other CELF4 SIRNA Assays ›
Gene Symbol:
CELF4
Gene Name:
CUGBP, Elav-like family member 4
Gene Aliases:
BRUNOL4, CELF-4
Chromosome Location:
Chr.18: 37243040 - 37566052 on Build GRCh38
UniGene ID:
Hs.435976
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID
289834
Purification
HPLC
Standard
In-Vivo Ready
Size
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
Availability
Inventoried
Catalog #
AM16708
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: CELF4
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_001025087.1
NP_001020258.1
13
2337
NM_001025088.1
NP_001020259.1
13
2334
NM_001025089.1
NP_001020260.1
12
2226
NM_020180.3
NP_064565.1
13
2340
XM_005258308.3
XP_005258365.1
13
2346
XM_011526079.2
XP_011524381.1
13
2414
XM_011526081.2
XP_011524383.1
13
2248
XM_011526083.2
XP_011524385.1
13
2411
XM_011526084.2
XP_011524386.1
13
2408
XM_011526085.2
XP_011524387.1
13
2409
XM_011526086.2
XP_011524388.1
13
2380
XM_011526087.2
XP_011524389.1
13
2379
XM_011526090.2
XP_011524392.1
13
2349
XM_011526091.2
XP_011524393.1
12
2268
XM_011526092.2
XP_011524394.1
12
2265
XM_017025840.1
XP_016881329.1
13
2247
XM_017025841.1
XP_016881330.1
13
2348
XM_017025842.1
XP_016881331.1
13
2350
XM_017025843.1
XP_016881332.1
13
2347
XM_017025844.1
XP_016881333.1
13
2347
XM_017025845.1
XP_016881334.1
13
2344
XM_017025846.1
XP_016881335.1
13
2321
XM_017025847.1
XP_016881336.1
13
2318
XM_017025848.1
XP_016881337.1
13
2316
XM_017025849.1
XP_016881338.1
13
2313
XM_017025850.1
XP_016881339.1
13
2184
XM_017025851.1
XP_016881340.1
13
2181
XM_017025852.1
XP_016881341.1
13
2181
XM_017025853.1
XP_016881342.1
12
2266
XM_017025854.1
XP_016881343.1
12
2266
XM_017025855.1
XP_016881344.1
12
2263
XM_017025856.1
XP_016881345.1
12
2263
XM_017025858.1
XP_016881347.1
12
2101
XM_017025859.1
XP_016881348.1
12
2098
XM_017025860.1
XP_016881349.1
12
2235
XM_017025861.1
XP_016881350.1
12
2100
XM_017025862.1
XP_016881351.1
12
2100
XM_017025863.1
XP_016881352.1
12
2079
XM_017025864.1
XP_016881353.1
12
2071
XM_017025865.1
XP_016881354.1
12
2097
XM_017025866.1
XP_016881355.1
13
1772
XM_017025867.1
XP_016881356.1
13
1746
XM_017025868.1
XP_016881357.1
13
1722
XM_017025869.1
XP_016881358.1
13
1713
XM_017025870.1
XP_016881359.1
13
1719
XM_017025871.1
XP_016881360.1
12
1638
XM_017025872.1
XP_016881361.1
12
1629
GenBank mRNA
AK126716.1
-
10
3091
BC001946.1
-
13
2095
2820541
BC004167.2
-
13
2094
2820541
BC045711.1
-
12
1913
5261155
Back To Top
More Information
Gene Ontology Categories:
Function(s)
Process(es)
alternative mRNA splicing, via spliceosome
regulation of alternative mRNA splicing, via spliceosome
mRNA splice site selection
germ cell development
embryo development
negative regulation of translation
negative regulation of mRNA splicing, via spliceosome
positive regulation of mRNA splicing, via spliceosome
negative regulation of excitatory postsynaptic potential
nucleotide binding
translation repressor activity, nucleic acid binding
mRNA binding
BRE binding
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-blue-65d5d77f49-g5d4h:80/100.66.76.71:80.
git-commit: a4c025c8ba47546866237b65de1f276a5fdbbe78
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.18.1-Offline