Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: COQ8A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_020247.4 | NP_064632.2 | 6, 7 | 1032 | ||
XM_005273201.1 | XP_005273258.1 | 6, 7 | 928 | |||
XM_011544238.1 | XP_011542540.1 | 7, 8 | 1124 | |||
XM_011544239.2 | XP_011542541.1 | 8, 9 | 1147 | |||
XM_011544240.2 | XP_011542542.1 | 6, 7 | 941 | |||
XM_011544241.2 | XP_011542543.1 | 6, 7 | 903 | |||
XM_017001852.1 | XP_016857341.1 | 6, 7 | 1122 | |||
GenBank mRNA | AB073905.1 | - | 6, 7 | 955 | ||
AJ278126.1 | - | 2, 3 | 250 | |||
AK090494.1 | - | 6, 7 | 769 | |||
AK092784.1 | - | 6, 7 | 755 | |||
AK293571.1 | - | 7, 8 | 929 | |||
AK297735.1 | - | 3, 4 | 311 | |||
AK300537.1 | - | 3, 4 | 262 | |||
AK301012.1 | - | 8, 9 | 953 | |||
BC005171.2 | - | 6, 7 | 981 | 3604948 | ||
BC129930.1 | - | 12, 13 | 1874 | 40010265 | ||
BX648860.1 | - | 6, 7 | 853 |