Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: HACE1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001321080.1 | NP_001308009.1 | 23 | 3353 | ||
NM_001321083.1 | NP_001308012.1 | 24 | 3300 | |||
NM_001321084.1 | NP_001308013.1 | 23 | 3217 | |||
NM_020771.3 | NP_065822.2 | 24 | 3485 | |||
NR_104424.1 | 26 | 3684 | ||||
XM_017011113.1 | XP_016866602.1 | 24 | 3352 | |||
XM_017011114.1 | XP_016866603.1 | 23 | 3171 | |||
XM_017011115.1 | XP_016866604.1 | 23 | 3327 | |||
XM_017011116.1 | XP_016866605.1 | 23 | 3382 | |||
XM_017011118.1 | XP_016866607.1 | 21 | 2905 | |||
XM_017011119.1 | XP_016866608.1 | 23 | 3188 | |||
XM_017011120.1 | XP_016866609.1 | 22 | 3303 | |||
XM_017011121.1 | XP_016866610.1 | 22 | 3270 | |||
XM_017011122.1 | XP_016866611.1 | 23 | 3022 | |||
GenBank mRNA | AB037741.1 | - | 13 | 4232 | ||
AJ420474.1 | - | 2 | 662 | |||
AK124268.1 | - | 2 | 2647 | |||
AK131207.1 | - | 19 | 2841 | |||
AL834202.1 | - | 11 | 1722 | |||
BC034982.2 | - | 24 | 3449 | 4838835 |