Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MIB1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_020774.3 | NP_065825.1 | 12 | 1986 | ||
XM_011526098.1 | XP_011524400.1 | 4 | 987 | |||
XM_017025873.1 | XP_016881362.1 | 9 | 1678 | |||
GenBank mRNA | AK091266.1 | - | 11 | 1416 | ||
AY147849.1 | - | 12 | 2004 | |||
AY149908.1 | - | 12 | 1731 | |||
BC022403.1 | - | 2 | 107 | |||
BC110581.1 | - | 12 | 1731 | 40008394 | ||
BC110582.1 | - | 12 | 1731 | 40008395 | ||
CR749635.1 | - | 9 | 1151 |