Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ZNF624 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_020787.3 | NP_065838.2 | 6 | 687 | ||
XM_006721562.3 | XP_006721625.1 | 6 | 868 | |||
XM_011523970.2 | XP_011522272.1 | 6 | 2038 | |||
GenBank mRNA | AB037770.1 | - | 1 | 508 | ||
AK131401.1 | - | 6 | 678 | |||
BC103943.1 | - | 1 | 269 | 40008882 | ||
BC103944.1 | - | 1 | 269 | 40008883 | ||
BC103946.1 | - | 1 | 269 | 40008885 |