Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: STAMBPL1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_020799.3 | NP_065850.1 | 9 | 1586 | ||
XM_006717928.2 | XP_006717991.1 | 10 | 1687 | |||
XM_006717930.3 | XP_006717993.1 | 9 | 2174 | |||
XM_011539983.1 | XP_011538285.1 | 10 | 2094 | |||
XM_011539985.1 | XP_011538287.1 | 10 | 1417 | |||
XM_017016453.1 | XP_016871942.1 | 10 | 2411 | |||
XM_017016454.1 | XP_016871943.1 | 11 | 1895 | |||
XM_017016455.1 | XP_016871944.1 | 12 | 2350 | |||
XM_017016456.1 | XP_016871945.1 | 10 | 2007 | |||
XM_017016457.1 | XP_016871946.1 | 8 | 1795 | |||
XM_017016458.1 | XP_016871947.1 | 7 | 1111 | |||
GenBank mRNA | AB010120.1 | - | 8 | 1114 | ||
AB037794.1 | - | 9 | 1907 | |||
AK056086.1 | - | 9 | 1515 | |||
AK090678.1 | - | 4 | 1407 | |||
AK092946.1 | - | 8 | 1715 | |||
BC010846.1 | - | 9 | 1504 | 3898533 | ||
BX649096.1 | - | 10 | 10009 |