Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CNTN3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_020872.2 | NP_065923.1 | 22 | 3066 | ||
XM_005264757.3 | XP_005264814.1 | 23 | 3176 | |||
XM_011533768.2 | XP_011532070.1 | 22 | 6698 | |||
XM_017006507.1 | XP_016861996.1 | 23 | 3746 | |||
XM_017006508.1 | XP_016861997.1 | 22 | 2971 | |||
XM_017006509.1 | XP_016861998.1 | 13 | 1745 | |||
GenBank mRNA | AB040929.1 | - | 20 | 2715 | ||
AL512746.1 | - | 2 | 91 | |||
BC150608.1 | - | 22 | 3066 | 9056978 |