Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: DHRS4L2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK021587.1 | - | 1 | 689 | ||
GU564234.1 | - | 7 | 566 | |||
GU564235.1 | - | 6 | 668 |
Gene Symbol: DHRS4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001282988.1 | NP_001269917.1 | 5 | 522 | ||
NM_021004.3 | NP_066284.2 | 6 | 624 | |||
GenBank mRNA | AB045131.1 | - | 6 | 649 | ||
AF044127.1 | - | 6 | 595 | |||
AF064256.1 | - | 6 | 598 | |||
AK001870.1 | - | 6 | 590 | |||
AK314448.1 | - | 6 | 589 | |||
AY358638.1 | - | 6 | 605 | |||
BC003019.1 | - | 6 | 572 | 2822884 | ||
DQ325464.1 | - | 5 | 489 | |||
DQ344810.1 | - | 6 | 603 |