Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: RBM25 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_021239.2 | NP_067062.1 | 4 | 405 | ||
XM_011537044.2 | XP_011535346.1 | 5 | 507 | |||
XM_011537045.2 | XP_011535347.1 | 3 | 212 | |||
GenBank mRNA | AK026107.1 | - | 4 | 385 | ||
AK094697.1 | - | 3 | 202 | |||
AL832314.1 | - | 4 | 376 | |||
BC062440.1 | - | 4 | 409 | |||
BC083496.1 | - | 4 | 371 | |||
BC111066.1 | - | 4 | 369 | |||
BC113389.1 | - | 3 | 185 | |||
BC113391.1 | - | 3 | 185 | |||
BC136775.1 | - | 4 | 242 | 9020765 | ||
BC136776.1 | - | 4 | 242 | 9020766 | ||
BC144407.1 | - | 5 | 310 | 9052928 | ||
BX647116.1 | - | 4 | 358 |