Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SIVA1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_006427.3 | NP_006418.2 | 3 | 412 | ||
NM_021709.2 | NP_068355.1 | 2 | 217 | |||
XM_011536360.2 | XP_011534662.1 | 3 | 1125 | |||
GenBank mRNA | AF033111.1 | - | 3 | 238 | ||
AK128704.1 | - | 2 | 1122 | |||
AK223332.1 | - | 3 | 398 | |||
AK300415.1 | - | 3 | 361 | |||
AK316457.1 | - | 3 | 343 | |||
AK316550.1 | - | 3 | 398 | |||
BC034562.1 | - | 2 | 191 | 3860364 | ||
BU195069.1 | - | 3 | 412 | |||
CK001877.1 | - | 3 | 406 |