Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CYP4F12 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_023944.3 | NP_076433.3 | 13 | 1711 | ||
NR_117085.1 | 11 | 1933 | ||||
XM_006722850.3 | XP_006722913.1 | 13 | 1596 | |||
XM_011528202.2 | XP_011526504.1 | 11 | 1231 | |||
XM_011528203.2 | XP_011526505.1 | 10 | 2453 | |||
XM_011528204.2 | XP_011526506.1 | 11 | 1932 | |||
XM_011528205.2 | XP_011526507.1 | 10 | 1144 | |||
XM_011528207.2 | XP_011526509.1 | 10 | 2840 | |||
XM_011528208.2 | XP_011526510.1 | 12 | 1350 | |||
XM_017027172.1 | XP_016882661.1 | 11 | 1454 | |||
GenBank mRNA | AB035130.1 | - | 13 | 1622 | ||
AB035131.1 | - | 13 | 1622 | |||
AK074412.1 | - | 6 | 3217 | |||
AK075435.1 | - | 11 | 1861 | |||
AL832171.1 | - | 11 | 2822 | |||
AY358977.1 | - | 13 | 1633 | |||
DR422336.1 | - | 2 | 368 |