Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: VAT1L | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_020927.2 | NP_065978.1 | 9 | 3652 | ||
GenBank mRNA | AB046796.1 | - | 9 | 3622 | ||
AK001256.1 | - | 1 | 1986 | |||
AL834240.1 | - | 8 | 3267 | |||
BC033537.1 | - | 9 | 3630 | 4821265 |