Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: HUWE1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_031407.6 | NP_113584.3 | 11 | 1154 | ||
XM_005261965.3 | XP_005262022.1 | 10 | 1221 | |||
XM_011530747.1 | XP_011529049.1 | 11 | 1148 | |||
XM_011530751.2 | XP_011529053.1 | 10 | 1016 | |||
XM_017029191.1 | XP_016884680.1 | 9 | 3879 | |||
XM_017029192.1 | XP_016884681.1 | 9 | 3879 | |||
XM_017029193.1 | XP_016884682.1 | 9 | 3879 | |||
XM_017029194.1 | XP_016884683.1 | 9 | 3879 | |||
XM_017029195.1 | XP_016884684.1 | 9 | 3879 | |||
XM_017029196.1 | XP_016884685.1 | 9 | 3879 | |||
XM_017029197.1 | XP_016884686.1 | 9 | 3879 | |||
XM_017029198.1 | XP_016884687.1 | 10 | 1221 | |||
XM_017029199.1 | XP_016884688.1 | 11 | 1148 | |||
XM_017029200.1 | XP_016884689.1 | 10 | 1018 | |||
XM_017029201.1 | XP_016884690.1 | 10 | 1035 | |||
XM_017029202.1 | XP_016884691.1 | 10 | 1011 | |||
XM_017029203.1 | XP_016884692.1 | 10 | 1017 | |||
XM_017029204.1 | XP_016884693.1 | 9 | 3878 | |||
XM_017029205.1 | XP_016884694.1 | 9 | 3879 | |||
XM_017029206.1 | XP_016884695.1 | 9 | 3879 | |||
GenBank mRNA | AY772009.1 | - | 9 | 799 | ||
AY929612.1 | - | 8 | 751 | |||
DQ097177.1 | - | 11 | 1153 |