Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: COQ5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_032314.3 | NP_115690.3 | 3 | 541 | ||
XM_006719639.2 | XP_006719702.1 | 4 | 641 | |||
GenBank mRNA | AK057777.1 | - | 3 | 375 | ||
AK222610.1 | - | 3 | 531 | |||
AK225242.1 | - | 3 | 531 | |||
AK293656.1 | - | 3 | 562 | |||
BC004916.2 | - | 3 | 517 | |||
BC107874.1 | - | 3 | 535 | 6452052 | ||
BP307378.1 | - | 3 | 541 | |||
BX647562.1 | - | 3 | 530 | |||
EU700459.1 | - | 3 | 535 |